missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ TC-treated Multiwell Variety Packs

Choose from different groups of cellware conveniently packaged together in popular combinations

176.55 € - 202.23 €

Specifications

Type Cellware Variety Pack
Storage Requirements 2° to 8°C
View More Specs

Products 2
Product Code Brand Item Description Price Quantity & Availability  
Product Code Brand Item Description Price Quantity & Availability  
10586571
View Documents Promotion Details
Corning™
354417
Variety pack I This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10335631
View Documents Promotion Details
Corning™
354431
Variety pack II This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Selections include different combinations of extracellular matrix proteins and attachment factors

Multiwell Plate Variety Packs

  • 6-well
  • Pack I contains five plates (clear, round well, flat-bottom, TC-treated, with 9.6cm2 cell growth area)
  • Coated with one of the following: Collagen I, Collagen IV, Fibronectin, Laminin, and Poly-D-Lysine
  • Lids (Fisher P/N 08-774-133 only)

Specifications

Specifications

Cellware Variety Pack
2° to 8°C
Videos
SDS
Documents

Documents

Product Certifications