Learn More
Corning™ BioCoat™ TC-treated Cultureslide Variety Pack
Choose from different groups of cellware conveniently packaged together in popular combinations
Brand:  Corning™ 354656
Product Code. 10498842
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Selections include different combinations of extracellular matrix proteins and attachment factors
Cultureslide Variety Packs
  • 8-well
  • Pack contains twelve slides coated with Collagen I, Fibronectic and Poly-D-Lysine
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Culture Slide
2° to 8°C