missing translation for 'onlineSavingsMsg'
Learn More

Copper turnings, 99+% (metals basis), Thermo Scientific Chemicals

20.40 € - 141.00 €

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381218
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010161.18
50 g
20.40 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11391218
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010161.30
250 g
50.00 €
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11301228
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010161.A1
1 kg
141.00 €
1kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-50-8
63.55
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]

Specifications

7440-50-8
99+% (Metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
63.546
0 hPa
Copper
Copper colored strands
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
63.55
CHEBI:30052
Odorless
Long, Thin, Coiled Strands
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-159-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only