missing translation for 'onlineSavingsMsg'
Learn More

Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis), Thermo Scientific Chemicals

46.03 € - 489.44 €

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11331509
SDS View Documents
Thermo Scientific Alfa Aesar
013990.18
50 g
46.03 €
50g
Please to purchase this item. Need a web account? Register with us today!
11301509
SDS View Documents
Thermo Scientific Alfa Aesar
013990.36
500 g
64.62 €
500g
Please to purchase this item. Need a web account? Register with us today!
11311509
SDS View Documents
Thermo Scientific Alfa Aesar
013990.A3
2 kg
135.68 €
2kg
Please to purchase this item. Need a web account? Register with us today!
11321509
SDS View Documents
Thermo Scientific Alfa Aesar
013990.A9
10 kg
489.44 €
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-50-8
63.55
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]

Specifications

7440-50-8
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
63.55
CHEBI:30052
−325 Mesh 10% max +325 Mesh
APS 8 to 11 Micron (FSSS)
(metals basis)
Cu
UN3089
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
99%
Odorless
Powder
Copper powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335-H500

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

missing translation for 'einecsNumber' : 231-159-6

missing translation for 'rtecsNumber' : GL5325000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.