missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) Sulfate Pentahydrate, Extra Pure, SLR, Fisher Chemical™
26.25 € - 218.00 €
Chemical Identifiers
CAS | 7758-99-8 |
---|---|
Molecular Formula | CuH10O9S |
Molecular Weight (g/mol) | 249.68 |
MDL Number | 149681 |
InChI Key | JZCCFEFSEZPSOG-UHFFFAOYSA-L |
IUPAC Name | copper(2+) pentahydrate sulfate |
SMILES | O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10368193
|
Fisher Chemical
C/8520/50 |
250 g | N/A | ||||||
10426870
|
Fisher Chemical
C/8520/53 |
500 g | N/A | ||||||
10743791
|
Fisher Chemical
C/8520/60 |
1 kg |
69.45 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10519380
|
Fisher Chemical
C/8520/63 |
3 kg | N/A | ||||||
10010090
|
Fisher Chemical
C/8520/65 |
5 kg | N/A | ||||||
Description
Copper(II) Sulfate Pentahydrate, Extra Pure, SLR
Chemical Identifiers
7758-99-8 | |
249.68 | |
JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
CuH10O9S | |
149681 | |
copper(2+) pentahydrate sulfate |
Specifications
110°C | |
Blue | |
Solid | |
149681 | |
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O | |
249.68 | |
Plastic powder jar |
7758-99-8 | |
3.5 to 4.5 | |
CuH10O9S | |
JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
copper(2+) pentahydrate sulfate | |
249.68g/mol | |
7.3mmHg 25 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- COPPER (II) SULFATE PENTAHYDRATE
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : Copper (II) sulfatepentahydrate (1:1:5), Cupric sulfate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title