missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate, 98%, pure, anhydrous, Thermo Scientific Chemicals

25.02 € - 331.65 €

Chemical Identifiers

CAS 7758-98-7
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.6
MDL Number MFCD00010981
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
PubChem CID 24462
ChEBI CHEBI:23414
IUPAC Name copper;sulfate
SMILES [O-]S(=O)(=O)[O-].[Cu+2]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10341451
View Documents
Thermo Scientific Acros
422870050
5 g Glass bottle
25.02 €
5g
Please to purchase this item. Need a web account? Register with us today!
10413465
View Documents
Thermo Scientific Acros
422871000
100 g Glass bottle
31.96 €
100g
Please to purchase this item. Need a web account? Register with us today!
10781062
View Documents
Thermo Scientific Acros
422875000
500 g Glass bottle
94.47 €
500g
Please to purchase this item. Need a web account? Register with us today!
11904241
View Documents
Thermo Scientific Acros
422870025
2.5 kg Glass bottle
331.65 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7758-98-7
159.6
ARUVKPQLZAKDPS-UHFFFAOYSA-L
24462
copper;sulfate
CuO4S
MFCD00010981
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
CHEBI:23414
[O-]S(=O)(=O)[O-].[Cu+2]

Specifications

200.0°C
Gray
97.5% min. (Iodometry)
CuSO4
01,164; 02,89; 05,162; 06,141; 08,125; 10,107; 16,283
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
ARUVKPQLZAKDPS-UHFFFAOYSA-L
copper;sulfate
24462
159.6
Pure
7758-98-7
Powder
CuO4S
MFCD00010981
15,2643
Solubility in water: 203g/L (20°C).
[O-]S(=O)(=O)[O-].[Cu+2]
159.6
CHEBI:23414
98%
Copper(II) sulfate, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if y

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 231-847-6

Product Identifier
  • Copper(II) sulfate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only