missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Chloride, Honeywell™

Product Code. 15661950
Click to view available options
Quantity:
25 g
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-89-6
Molecular Formula ClCu
Molecular Weight (g/mol) 98.996
MDL Number MFCD00010971
InChI Key OXBLHERUFWYNTN-UHFFFAOYSA-M
Synonym cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
PubChem CID 62652
ChEBI CHEBI:53472
IUPAC Name chlorocopper
SMILES Cl[Cu]

Specifications

Quantity 500 g
Linear Formula CuCl
UN Number 2802
Solubility Information Partially soluble in dimethyl sulfide, Insoluble in water
Formula Weight 99g/mol
Percent Purity 97%
Grade Reagent
Vapor Pressure 1.3mmHg (546°C)
Chemical Name or Material Copper(I) Chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'