missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Granular, 20 to 30 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
Specifications
Quantity | 500 g |
Formula Weight | 63.55 |
Grade | BAKER ANALYZED™ Reagent |
Mesh Size | 20 to 30 Mesh |
Packaging | Poly Bottle |
Form | Granular |
Chemical Name or Material | Copper |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'