Learn More
Contec™ CONSTIX SP-6N Sealed Polyester Swabs
BIOPHARMA
Contec™ CONSTIX SP-6N Sealed Polyester Swabs consists of a polyester nonwoven fabric that is thermally bonded to a flex-tip polypropylene handle.
Brand:  Contec™ SP-6N
Product Code. 15835204
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Flexible tip provides a softer application on surfaces
  • Nonwoven polyester head provides enhanced sorbency for non-aqueous solutions
Specifications
Specifications
3.0 mm
16.3 mm
Non-sterile
Polypropylene
2.5 mm
162.6 mm
6.9 mm