Learn More
DWK Life Sciences DURAN™ Stainless Steel Multiport GL 45 Connector Cap, from PBT, GL 45, with 2 Ports
Description
A robust and durable tubing connection system for use with all sizes of DURAN™ GL 45 laboratory and media bottles. The bottle top adaptor facilitates the transfer of liquids within a closed and sterile system. The barbed hose connectors are suitable for flexible tubing with an 8.0 mm inner diameter. Freely rotating Stainless steel center allows the bottle to be unscrewed without the need to disconnect the tubing. Connectors and body are manufactured from Type 316L stainless steel. A silicone gasket and the red PBT GL 45 screw cap provide a liquid tight seal. Wide temperature usage range (up to 180°C). Autoclavable (121°C for 15 minutes). Retrace coded for full traceability.
- Facilitated transfer of liquids within a closed and sterile system due to the bottle top adaptor
- The one, two or three barbed, male hose connectors on both the top and bottom assemblies are suitable for connecting flexible Silicone tubing with a 8.0 mm inner diameter
- Freely rotating Stainless steel center allows the bottle to be unscrewed without the need to disconnect the tubing
- Easier attachment of the tubing due to slightly splayed top connectors
- All components are fully welded to avoid entrapment voids
- Hygienic, electropolished surface for efficient cleaning and sterilisation
- A USP <88> Class VI silicone gasket and the red PBT GL 45 screw cap provide a liquid tight seal
- Lot coded for tracability
Specifications
Specifications
For Use With (Application) | Sterile fluid transfer in and out of lab bottles; Supply of reagents to instruments; Port assemblies for spinner flasks, and cell culture vessels; Autoclaving of media in bottles |
Quantity | Case of 1 Connector Cap |