missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 1413μS/cm, NIST Certified, Chem-Lab
Calibrates electrodes and meters for accurate electrical and thermal conductivity measurements. Conductivity Standard, 1413μS/cm, NIST Certified, Chem-Lab is a 0.01M potassium chloride solution for laboratory use and LF measurements. 1278μS at 20°C, 1413μS at 25°C (±1%).
Chemical Identifiers
CAS | 7447-40-7 |
---|---|
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.55 |
MDL Number | MFCD00011360 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
SMILES | [Cl-].[K+] |
Description
5°C—896μS/cm
10°C—1020μS/cm
15°C—1147μS/cm
20°C—1278μS/cm
25°C—1413μS/cm
30°C—1552μS/cm
35°C—1694μS/cm
40°C—1839μS/cm
Chemical Identifiers
7447-40-7 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
CHEBI:32588 |
ClK | |
MFCD00011360 | |
4873 | |
[Cl-].[K+] |
Specifications
7447-40-7 | |
1.01g/mL | |
ClK | |
0.01 M | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 | |
Potassium Chloride |
Polyethylene | |
1413μS/cm | |
MFCD00011360 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium;chloride | |
4873 | |
74.56g/mol |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Conductivity Standard, 1413μS/cm, NIST Certified, Chem-Lab