missing translation for 'onlineSavingsMsg'
Learn More

1413 μS/cm Conductivity Standard Solution Sachets, Hanna Instruments™

Premium quality 1,413 μS/cm conductivity solution that is NIST traceable and supplied with a Certificate of Analysis that is ideal for users that do not calibrate frequently.

Specifications

Color Colorless
Odor Odorless
pH 7
Solubility Soluble
Physical Form Liquid
View More Specs

Description

Description

  • Premium quality 1,413 μS/cm conductivity solution that is NIST traceable and supplied with a Certificate of Analysis
  • Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
  • 1,413 μS/cm conductivity solution standardized with a conductivity meter that has been calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material)  
  • Deionized water for analytical use in accordance with ISO 3696/BS3978
  • Additional equipment used in preparation of the solutions include certified weight-checked balances, Class A glassware and certified thermometers
  • Ideal for users that do not calibrate frequently
  • Ideal for field use due to portability
Specifications

Specifications

Colorless
7
Liquid
Odorless
Soluble
Videos
SDS
Documents

Documents

Product Certifications