Learn More
Cole-Parmer™ Compression to Male Threaded Adapter
Compression fittings are an excellent choice for connecting rigid tubing systems
Brand:  Cole-Parmer™ 06385-87
 View more versions of this product
Product Code. 11749539
  • 18.15 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
  • PVDF Composition
Specifications
Specifications
Compression to Male Threaded Adapter
Autoclavable
1 5/8 in.
1/2 in.
1/2 in. NPT(M)
24 mm
Straight
Compression x NPT(M) Connection
PVDF
10 Fittings
1/2 in.
Autoclave or Ethylene Oxide
15/16 in.