missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Comacier VHP Plus Heavy Duty Leather Gloves

Provides protection against cuts with flexibility and comfort. Ansell Comacier VHP Plus Heavy Duty Leather Gloves are made from high performance fibres using Techcor™ Knitting Technology.

243.00 €

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2003
Color Gray
Exterior Finish Leather Palm
Material Leather
Product Type Mechanical Protection Gloves
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15535746
View Documents
Ansell™
97039100
10 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15545746
View Documents
Ansell™
97039110
11 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15555746
View Documents
Ansell™
97039080
8 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15565746
View Documents
Ansell™
97039090
9 243.00 €
12 pairs
Qty Price Break Available
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Mechanical protection heavy duty gloves
  • Medium weight 10 gauge seamless knitted glove
  • Knit wrist ensures good fit
  • Unique traceability system on textile tag
  • Palm, fingers and artery guard made of leather for optimal protection
  • Provides the highest levels of mechanical resistance
  • Good flexibility and comfort
  • Superior grip when used with oils and greases due to leather patch

Applications:

  • Handling heavy and oily metal pieces
  • Handling sharp or irregular-edged metal parts
  • Press and stamping
Specifications

Specifications

EN 420:2003 + A1:2009 and EN 388:2003
Leather Palm
Mechanical Protection Gloves
Techcor™
VHP Plus
Gray
Leather
Knit
12 pairs in bag, 10 bags in carton
240 to 260 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.