Learn More
OHAUS™ Column Clamp
OHAUS™ Column Clamp is designed to securely grip and position a variety of laboratory glassware.
Brand:  OHAUS™ 30392316
 View more versions of this product
Product Code. 15702369
  • 73.25 € / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Corrosion-resistant stainless steel clamps provide excellent chemical resistance, durability, and can be autoclaved. Nickel-plated zinc clamps offer high-tensile strength at an economical price.
  • Prevent accidents in the lab! Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects. Select from a variety of grip sizes, prong styles and adjustment types.
  • Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Column Clamp
Clamp, Specialty, Column, CLS-COLMNSL
13.31 in.
338.00 mm
Large
Column
Stainless Steel
11 mm
8.310 in.
211 mm
64 to 140 mm