missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL14A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA554886
This item is not returnable.
View return policy
Description
Immunogen sequence: LLVTPTSGGK TNQLNLQNTA TKAIIQGLMP DQNYTVQIIA YNKDKESKPA QGQFRIKDLE KRKDPKPR Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
Type XIV collagen is a fibril-associated collagen with an interrupted triple helix. It interacts with the fibril surface and regulates fibrillogenesis (Ansorge et al., 2009 [PubMed 19136672]).Specifications
COL14A1 | |
Polyclonal | |
Unconjugated | |
COL14A1 | |
5730412L22Rik; AW108078; col14a1; Collagen alpha-1(XIV) chain; collagen type XIV alpha 1 chain; collagen, type XIV, alpha 1; collagen, type XIV, alpha 1 (undulin); procollagen, type XIV, alpha 1; UND; Undulin; undulin (fibronectin-tenascin-related) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7373 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q05707 | |
COL14A1 | |
Recombinant protein corresponding to Human COL14A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only