missing translation for 'onlineSavingsMsg'
Learn More

Cobalt slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11309308
Click to view available options
Quantity:
1 Slug
25 Slugs
5 Slugs
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Quantity 25 Slugs
Percent Purity 99.995%
Grade Puratronic™
Odor Odorless
Weight ≈3.6g/slug
Form Slug
Assay (metals basis)
Chemical Name or Material Cobalt slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.