missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfide, 99.5% (metals basis), Thermo Scientific Chemicals

89.85 € - 313.56 €

Chemical Identifiers

CAS 1317-42-6
Molecular Formula CoS
Molecular Weight (g/mol) 90.99
MDL Number MFCD00016032
InChI Key INPLXZPZQSLHBR-UHFFFAOYSA-N
Synonym cobalt sulfide, sycoporite, cobalt ii sulfide, cobalt sulphide, cobaltous sulfide, cobalt 2+ sulfide, cobalt monosulfide, cobalt sulfide amorphous, ccris 7728
PubChem CID 14832
IUPAC Name sulfanylidenecobalt
SMILES [S--].[Co++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11332208
SDS View Documents
Thermo Scientific Alfa Aesar
013114.14
25 g
89.85 €
25g
Please to purchase this item. Need a web account? Register with us today!
11322208
SDS View Documents
Thermo Scientific Alfa Aesar
013114.22
100 g
313.56 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used in casting, smelting of metal, hard alloys. Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350

missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III

missing translation for 'einecsNumber' : 215-273-3

missing translation for 'rtecsNumber' : GG3325000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt(II) sulfide, 99.5% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.