Learn More
Cobalt(II) phosphate octahydrate, Thermo Scientific Chemicals
98.40 € - 359.00 €
Chemische Identifikatoren
CAS | 10294-50-5 |
---|---|
Molecular Formula | Co3H16O16P2 |
Molecular Weight (g/mol) | 510.859 |
MDL Number | MFCD00016028 |
InChI Key | QOQSIXJUYVUEMP-UHFFFAOYSA-H |
Synonym | cobalt ii phosphate octahydrate, cobaltous phosphate octahydrate, cobalt phosphate octahydrate, phosphoric acid, cobalt 2+ salt 2:3 , octahydrate, acmc-20ale3, 3co.2po4.8h2o, cobalt 2+ diphosphate octahydrate |
PubChem CID | 202398 |
IUPAC Name | cobalt(2+);diphosphate;octahydrate |
SMILES | O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2] |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
11302608
|
Thermo Scientific Alfa Aesar
023111.18 |
50 g |
98.40 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11392598
|
Thermo Scientific Alfa Aesar
023111.30 |
250 g |
359.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
It is used as a pigment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
10294-50-5 | |
510.859 | |
QOQSIXJUYVUEMP-UHFFFAOYSA-H | |
202398 | |
O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2] |
Co3H16O16P2 | |
MFCD00016028 | |
cobalt ii phosphate octahydrate, cobaltous phosphate octahydrate, cobalt phosphate octahydrate, phosphoric acid, cobalt 2+ salt 2:3 , octahydrate, acmc-20ale3, 3co.2po4.8h2o, cobalt 2+ diphosphate octahydrate | |
cobalt(2+);diphosphate;octahydrate |
Spezifikation
10294-50-5 | |
Co3H16O16P2 | |
cobalt ii phosphate octahydrate, cobaltous phosphate octahydrate, cobalt phosphate octahydrate, phosphoric acid, cobalt 2+ salt 2:3 , octahydrate, acmc-20ale3, 3co.2po4.8h2o, cobalt 2+ diphosphate octahydrate | |
QOQSIXJUYVUEMP-UHFFFAOYSA-H | |
cobalt(2+);diphosphate;octahydrate | |
202398 | |
Odorless | |
Cobalt(II) phosphate octahydrate |
Powder | |
MFCD00016028 | |
Slightly soluble in water. Soluble in mineral acids. | |
O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2] | |
510.859 | |
510.87 (366.74 Anhydrous) | |
2.769 g/mL |
Sicherheit und Handhabung
GHS H Statement
H301-H341-H317
Toxic if swallowed.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
H302-H315-H317-H319-H334-H335-H350
missing translation for 'einecsNumber' : 236-655-6
missing translation for 'rtecsNumber' : TB9279500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only