missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) phosphate, anhydrous, 98%, Thermo Scientific Chemicals

40.20 € - 123.62 €

Chemical Identifiers

CAS 13455-36-2
Molecular Formula Co3O8P2
Molecular Weight (g/mol) 366.739
MDL Number MFCD00016028
InChI Key ZBDSFTZNNQNSQM-UHFFFAOYSA-H
Synonym cobalt phosphate, cobaltous phosphate, cobalt ii phosphate, tricobalt bis orthophosphate, unii-0b5m38t47h, cobalt ii phosphate, anhydrous, phosphoric acid, cobalt 2+ salt 2:3, 3co.2po4, cobalt ii phosphate octahydrate, phosphoric acid,cobalt 2+ salt 2:3
PubChem CID 61615
IUPAC Name cobalt(2+);diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11479567
View Documents
Thermo Scientific Alfa Aesar
B22066.14
25 g
40.20 €
25g
Please to purchase this item. Need a web account? Register with us today!
11489567
View Documents
Thermo Scientific Alfa Aesar
B22066.22
100 g
123.62 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13455-36-2
366.739
ZBDSFTZNNQNSQM-UHFFFAOYSA-H
61615
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]
Co3O8P2
MFCD00016028
cobalt phosphate, cobaltous phosphate, cobalt ii phosphate, tricobalt bis orthophosphate, unii-0b5m38t47h, cobalt ii phosphate, anhydrous, phosphoric acid, cobalt 2+ salt 2:3, 3co.2po4, cobalt ii phosphate octahydrate, phosphoric acid,cobalt 2+ salt 2:3
cobalt(2+);diphosphate

Specifications

13455-36-2
MFCD00016028
ZBDSFTZNNQNSQM-UHFFFAOYSA-H
cobalt(2+);diphosphate
61615
98%
Cobalt(II) phosphate, anhydrous
Co3O8P2
cobalt phosphate, cobaltous phosphate, cobalt ii phosphate, tricobalt bis orthophosphate, unii-0b5m38t47h, cobalt ii phosphate, anhydrous, phosphoric acid, cobalt 2+ salt 2:3, 3co.2po4, cobalt ii phosphate octahydrate, phosphoric acid,cobalt 2+ salt 2:3
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]
366.739
366.74
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H350

missing translation for 'einecsNumber' : 236-655-6

missing translation for 'rtecsNumber' : TB9279000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.