missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals

45.43 € - 301.50 €

Chemical Identifiers

CAS 13478-33-6
Molecular Formula Cl2CoO8
Molecular Weight (g/mol) 257.825
MDL Number MFCD00150207
InChI Key BSUSEPIPTZNHMN-UHFFFAOYSA-L
Synonym cobaltous perchlorate, cobalt diperchlorate, cobalt ii perchlorate, cobalt diperchlorate hexahydrate, cobalt perchlorate hexahydrate, cobaltous perchlorate, hexahydrate, cobalt ii perchlorate, hexahydrate, perchloric acid, cobalt ii salt, hexahydrate, diperchloratocobalt
PubChem CID 26064
IUPAC Name cobalt(2+);diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11371758
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011652.09
10 g
45.43 €
10g
Please to purchase this item. Need a web account? Register with us today!
11351758
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011652.18
50 g
83.72 €
50g
Please to purchase this item. Need a web account? Register with us today!
11361758
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011652.30
250 g
301.50 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cobalt(II) perchlorate hexahydrate is used as chemical organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13478-33-6
257.825
BSUSEPIPTZNHMN-UHFFFAOYSA-L
26064
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2]
Cl2CoO8
MFCD00150207
cobaltous perchlorate, cobalt diperchlorate, cobalt ii perchlorate, cobalt diperchlorate hexahydrate, cobalt perchlorate hexahydrate, cobaltous perchlorate, hexahydrate, cobalt ii perchlorate, hexahydrate, perchloric acid, cobalt ii salt, hexahydrate, diperchloratocobalt
cobalt(2+);diperchlorate

Specifications

13478-33-6
Cl2CoO8
UN1481
Soluble in water.
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2]
257.825
366.74 (257.83 Anhydrous)
Odorless
Cobalt(II) perchlorate hexahydrate
Crystalline
MFCD00150207
cobaltous perchlorate, cobalt diperchlorate, cobalt ii perchlorate, cobalt diperchlorate hexahydrate, cobalt perchlorate hexahydrate, cobaltous perchlorate, hexahydrate, cobalt ii perchlorate, hexahydrate, perchloric acid, cobalt ii salt, hexahydrate, diperchloratocobalt
BSUSEPIPTZNHMN-UHFFFAOYSA-L
cobalt(2+);diperchlorate
26064
Reagent
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H351-H315-H319-H317-H335
May intensify fire
oxidizer.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P201-P202-P210-P220-P221-P261-P264b-P271-P280i-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P342+P311-P362-P370+P378q-P501c

H272-H315-H319-H334-H335-H350

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 236-653-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only