missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Nitrate Hexahydrate, Certified AR for Analysis, Fisher Chemical™

161.00 € - 180.00 €

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
IUPAC Name λ²-cobalt(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10529380
SDS View Documents
Fisher Chemical
C/6680/48
100 g
161.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
10000090
SDS View Documents
Fisher Chemical
C/6680/50
250 g
180.00 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

10026-22-9
291.03
QGUAJWGNOXCYJF-UHFFFAOYSA-N
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
CoH12N2O12
MFCD00149647
λ²-cobalt(2+) hexahydrate dinitrate

Specifications

55°C
Brown-Red
CoH12N2O12
QGUAJWGNOXCYJF-UHFFFAOYSA-N
λ²-cobalt(2+) hexahydrate dinitrate
291.02g/mol
10026-22-9
Solid
MFCD00149647
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
291.03
Plastic powder jar
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cobalt(II) nitrate hexahydrate

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Cobalt nitrate hexahydrate, Cobalt(II) nitrate

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt(II) Nitrate Hexahydrate, Certified AR for Analysis, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.