Learn More
Cobalt(II) hydroxide, 95%, pure, Thermo Scientific Chemicals
88.64 € - 339.69 €
Chemical Identifiers
CAS | 21041-93-0 |
---|---|
Molecular Formula | CoH2O2 |
Molecular Weight (g/mol) | 92.95 |
MDL Number | MFCD00016015 |
InChI Key | ASKVAEGIVYSGNY-UHFFFAOYSA-L |
Synonym | cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis |
PubChem CID | 10129900 |
SMILES | [OH-].[OH-].[Co++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15219846
|
Thermo Scientific Acros
453892500 |
250 g |
88.64 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15209846
|
Thermo Scientific Acros
453890010 |
1 kg |
339.69 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
21041-93-0 | |
92.95 | |
ASKVAEGIVYSGNY-UHFFFAOYSA-L | |
10129900 |
CoH2O2 | |
MFCD00016015 | |
cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis | |
[OH-].[OH-].[Co++] |
Specifications
21041-93-0 | |
CoH2O2 | |
MFCD00016015 | |
cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis | |
[OH-].[OH-].[Co++] | |
92.95 | |
92.95 | |
Pure |
95% | |
Co(OH)2 | |
15,2426 | |
ASKVAEGIVYSGNY-UHFFFAOYSA-L | |
λ²-cobalt(2+) dihydroxide | |
10129900 | |
95% | |
Cobalt(II) hydroxide |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Very toxic to aq
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 244-166-4
- Cobalt(II) hydroxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only