missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Chloride Hexahydrate, Technical, Fisher Chemical™

Product Code. 11428952
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Melting Point 86°C
Color Red to Violet
pH 4.6
Physical Form Crystalline Solid
Quantity 100 g
Formula Weight 237.93g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H341-Suspected of causing genetic defects.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/A§/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.