missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(Ii) Chloride Hexahydrate, ACS Reagent, Honeywell Fluka™
ACS Reagent, 98%
58.32 € - 386.19 €
Chemical Identifiers
CAS | 7791-13-1 |
---|---|
Molecular Formula | Cl2CoH12O6 |
Molecular Weight (g/mol) | 237.92 |
MDL Number | MFCD00149652 |
InChI Key | GFHNAMRJFCEERV-UHFFFAOYSA-L |
IUPAC Name | λ²-cobalt(2+) hexahydrate dichloride |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15642760
|
Honeywell-Fluka
255599-5G |
5 g |
58.32 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15622760
|
Honeywell-Fluka
255599-100G |
100 g |
95.56 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15632760
|
Honeywell-Fluka
255599-500G |
500 g |
386.19 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7791-13-1 | |
237.92 | |
GFHNAMRJFCEERV-UHFFFAOYSA-L | |
O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
Cl2CoH12O6 | |
MFCD00149652 | |
λ²-cobalt(2+) hexahydrate dichloride |
Specifications
7791-13-1 | |
CoCl2 · 6H2O | |
3077 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Co++] | |
237.92 | |
98% | |
Glass Bottle | |
Cobalt(Ii) Chloride Hexahydrate |
Cl2CoH12O6 | |
MFCD00149652 | |
GFHNAMRJFCEERV-UHFFFAOYSA-L | |
λ²-cobalt(2+) hexahydrate dichloride | |
237.93g/mol | |
ACS Reagent | |
40mmHg (0°C) |
Safety and Handling
P201-P261-P273-P280-P284-P304 + P340
H302-H317-H334-H341-H350i-H360F-H410
missing translation for 'einecsNumber' : 231-589-4
missing translation for 'rtecsNumber' : GG0200000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Cobalt(II) chloride hexahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : cobalt dichloride hexahydrate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title