missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride hexahydrate, 98%, Thermo Scientific Chemicals

30.75 € - 297.48 €

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobaltous chloride hexahydrate
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11437817
View Documents
Thermo Scientific Alfa Aesar
A16346.18
50 g
30.75 €
50g
Please to purchase this item. Need a web account? Register with us today!
11447817
View Documents
Thermo Scientific Alfa Aesar
A16346.30
250 g
95.17 €
250g
Please to purchase this item. Need a web account? Register with us today!
11427817
View Documents
Thermo Scientific Alfa Aesar
A16346.0B
1000 g
297.48 €
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cobalt(II) chloride hexahydrate is widely used as a drying agent in painting and coloring agents for glass ceramics. It is an excellent indicator for water in dessicants as well as an indicator for silica gel dryer. It helps in electroplating. It acts as an inducer of HIF-1 production used to study apoptotic effects in HepG2 cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) chloride hexahydrate is widely used as a drying agent in painting and coloring agents for glass ceramics. It is an excellent indicator for water in dessicants as well as an indicator for silica gel dryer. It helps in electroplating. It acts as an inducer of HIF-1 production used to study apoptotic effects in HepG2 cells.

Solubility
Soluble in water, ether, acetone and alcohol.

Notes
Incompatible with strong oxidizing agents and metals.
Specifications

Chemical Identifiers

7791-13-1
237.92
GFHNAMRJFCEERV-UHFFFAOYSA-L
λ²-cobalt(2+) hexahydrate dichloride
Cl2CoH12O6
MFCD00149652
Cobaltous chloride hexahydrate
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

87°C
4.9
CoCl2·6H2O
UN3077
Cobaltous chloride hexahydrate
GFHNAMRJFCEERV-UHFFFAOYSA-L
λ²-cobalt(2+) hexahydrate dichloride
237.93 (129.84 Anhydrous)
Odorless
1.924
7791-13-1
Cl2CoH12O6
MFCD00149652
14,2437
Soluble in water,ether,acetone and alcohol.
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
237.92
98%
Hygroscopic
Cobalt(II) chloride hexahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c

H302+H332-H317-H334-H341-H350i-H360F

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 231-589-4

missing translation for 'rtecsNumber' : GG0200000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Cobalt(II) chloride hexahydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.