Learn More
Cobalt(II) chloride, anhydrous, 99.7% (metals basis), Thermo Scientific Chemicals
Sensitivity: Hygroscopic; 3.367. Decomposes at 400°C on long heating in air
101.51 € - 391.95 €
Chemical Identifiers
CAS | 7646-79-9 |
---|---|
Molecular Formula | Cl2Co |
Molecular Weight (g/mol) | 129.83 |
MDL Number | MFCD00010938 |
InChI Key | GVPFVAHMJGGAJG-UHFFFAOYSA-L |
Synonym | cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride |
IUPAC Name | λ²-cobalt(2+) dichloride |
SMILES | [Cl-].[Cl-].[Co++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11361659
|
Thermo Scientific Alfa Aesar
012303.18 |
50 g |
101.51 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371659
|
Thermo Scientific Alfa Aesar
012303.30 |
250 g |
391.95 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cobalt(II) chloride is used in humidity indicator in weather instruments. In the anhydrous form, it finds use in electroplating of cobalt, in organic chemistry and is a precursor to cobaltocene, (bis(cyclopentadienyl)cobalt(II), which is a good reducing agent. It also serves as a Lewis acid. Cobalt chloride is an indicator for water in desiccants, owing to the reversible hydration/dehydration coupled with distinct color change. Cobalt chloride is useful for producing invisible ink as it turns blue when heated and becomes invisible once it gets cooled. Cobalt(II) chloride catalyzes cross coupling of aryl halides or vinyl halides with aryl grignard reagents in excellent yields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7646-79-9 | |
129.83 | |
GVPFVAHMJGGAJG-UHFFFAOYSA-L | |
λ²-cobalt(2+) dichloride |
Cl2Co | |
MFCD00010938 | |
cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride | |
[Cl-].[Cl-].[Co++] |
Specifications
1049°C | |
7646-79-9 | |
Powder | |
Cl2Co | |
cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride | |
GVPFVAHMJGGAJG-UHFFFAOYSA-L | |
λ²-cobalt(2+) dichloride | |
129.84 | |
Hygroscopic | |
Cobalt(II) chloride, Anhydrous |
735°C | |
Blue | |
99.7% (Metals basis) | |
MFCD00010938 | |
Freely soluble in water and methanol. Soluble in alcohol,acetone,ether,glycerol,and pyridine. | |
[Cl-].[Cl-].[Co++] | |
129.83 | |
Odorless | |
3.367 |
Safety and Handling
Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P363-P501c
H302+H332-H317-H318-H334-H335-H341-H350i-H360F-H373
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 231-589-4
missing translation for 'rtecsNumber' : GF9800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.