missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride, 0.5M aq. soln., Thermo Scientific™

Product Code. 15495509
Click to view available options
Quantity:
50 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Color Colorless
Physical Form Liquid
Quantity 50 mL
Linear Formula CoCl2
UN Number UN3082
Formula Weight 129.84
Concentration or Composition (by Analyte or Components) 0.5M aq. soln.
Chemical Name or Material Cobalt(II) chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'