missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(Ii) Acetate Tetrahydrate, Reagent grade, Honeywell™

Product Code. 15611840
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6147-53-1
Molecular Formula C4H14CoO8
Molecular Weight (g/mol) 249.081
MDL Number MFCD00149569
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
PubChem CID 61372
IUPAC Name cobalt(2+);diacetate;tetrahydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]

Specifications

Melting Point 298°C
Quantity 250 g
Linear Formula (CH3COO)2Co · 4H2O
UN Number UN3077
Beilstein 3731397
Formula Weight 249.08g/mol
Grade Reagent
Packaging Poly Bottle
Appearance Crystals
Density 1.705g/cu.cm
Chemical Name or Material Cobalt(Ii) Acetate Tetrahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'