missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(Ii) Acetate Tetrahydrate, Reagent grade, Honeywell™
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | 298°C |
Quantity | 1 kg |
Linear Formula | (CH3COO)2Co · 4H2O |
UN Number | UN3077 |
Beilstein | 3731397 |
Formula Weight | 249.08g/mol |
Grade | Reagent |
Packaging | Poly Bottle |
Appearance | Crystals |
Density | 1.705g/cu.cm |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'