missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt boride, 99%, Thermo Scientific™
71.00 € - 244.00 €
Chemical Identifiers
CAS | 12619-68-0 |
---|---|
Molecular Formula | B2Co3+6 |
Molecular Weight (g/mol) | 198.42 |
MDL Number | MFCD00146483 |
InChI Key | QSOMCIJHXYGMAO-UHFFFAOYSA-N |
Synonym | boron;cobalt 2+ |
PubChem CID | 121233614 |
IUPAC Name | boron;cobalt(2+) |
SMILES | [B].[B].[Co+2].[Co+2].[Co+2] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
12619-68-0 | |
198.42 | |
QSOMCIJHXYGMAO-UHFFFAOYSA-N | |
121233614 | |
[B].[B].[Co+2].[Co+2].[Co+2] |
B2Co3+6 | |
MFCD00146483 | |
boron;cobalt 2+ | |
boron;cobalt(2+) |
Specifications
12619-68-0 | |
B2Co3+6 | |
boron;cobalt 2+ | |
QSOMCIJHXYGMAO-UHFFFAOYSA-N | |
boron;cobalt(2+) | |
121233614 | |
Odorless |
Gray | |
MFCD00146483 | |
Insoluble in water. | |
[B].[B].[Co+2].[Co+2].[Co+2] | |
198.42 | |
99% | |
Cobalt boride |
Safety and Handling
GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350
missing translation for 'einecsNumber' : 235-722-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures