Learn More
Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers.
Brand:  Thermo Scientific™ FLN456LEV
 View more versions of this product
Product Code. 16367141
  • 3025.05 € / Each
Explore more special offers
Description
Description
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
Features:
  • LN2 backup system maintains temperature down to -80°C with liquid nitrogen
  • CO2 backup system maintains temperature down to -67°C with CO2
Ordering Information:
Available as field- or factory-installed. Models for 300 box capacity models are standalone. Field-installed units must be installed by qualified professionals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LN2 Backup System
Forma FDE, Revco RDE, HERAfreeze HDE, and Thermo Scientific TDE Series (400 box capacity and larger)
230 V
-80°C
LN2 Backup System: maintains temperature down to -80°C with LN2
Boxes (300)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers > ULT BUS; Type: LN2; Voltage: 230V/50Hz