Learn More
CNOT9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22503-1-AP-20UL
 View more versions of this product
Product Code. 16851955
  • 145.00 € / 20µL
Explore more special offers
Description
Description
This gene encodes a member of the highly conserved RCD1 protein family. The encoded protein is a transcriptional cofactor and a core protein of the CCR4-NOT complex. It may be involved in signal transduction as well as retinoic acid-regulated cell differentiation and development. Alternatively spliced transcript variants have been described for this gene.
Specifications
Specifications
CNOT9
Polyclonal
Unconjugated
CNOT9
CNOT9, Rcd 1, RCD1, RCD1+, RQCD1
Rabbit
Antigen Affinity Chromatography
RUO
301513, 58184, 9125
-20°C
Liquid
Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5PQL2, Q92600, Q9JKY0
Cnot9
CNOT9 Fusion Protein Ag18197
20 μL
Primary
Human, Rat, Mouse