Learn More
CNOT1 Mouse anti-Human, CoraLite 488, Clone: 1A10A11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66507-100UL
This item is not returnable.
View return policy
Description
The CNOT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CNOT1. This antibody recognizes human antigen. The CNOT1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
CNOT1 | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human |
Immunofluorescence | |
1A10A11 | |
CoraLite 488 | |
100 μL | |
23019 | |
IgG1 |