missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNKSR3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14084-1-AP-150UL
This item is not returnable.
View return policy
Description
CNKSR3 may be involved in transepithelial sodium transport. Regulates aldosterone-induced and ENaC-mediated sodium transport possibly through regulation of the ERK pathway.Specifications
CNKSR3 | |
Polyclonal | |
Unconjugated | |
CNKSR3 | |
CNK homolog protein 3, CNK3, CNKSR family member 3, CNKSR3, Connector enhancer of KSR 3, MAGI1, Maguin like protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
154043, 215748, 308113 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5SGD7, Q6P9H4, Q8BMA3 | |
CNKSR3 | |
CNKSR3 Fusion Protein Ag5213 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction