missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLRN2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23994-1-AP-150UL
This item is not returnable.
View return policy
Description
Clarin 2 belongs to the clarin family of genes. It′s a small integral membrane glycoproteins with four transmembrane domains.Specifications
CLRN2 | |
Polyclonal | |
Unconjugated | |
CLRN2 | |
clarin 2, CLRN2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
624224, 645104 | |
-20°C | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A0PK11 | |
Clrn2 | |
CLRN2 Fusion Protein Ag21182 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction