missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Clofazimine, 98%

61.61 € - 216.00 €

Chemical Identifiers

CAS 2030-63-9
Molecular Formula C27H22Cl2N4
Molecular Weight (g/mol) 473.4
InChI Key WDQPAMHFFCXSNU-UHFFFAOYSA-N
PubChem CID 2794
ChEBI CHEBI:3749
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15771779
View Documents
Thermo Scientific Chemicals
461760010
1 g
61.61 €
1g
Please to purchase this item. Need a web account? Register with us today!
15758817
View Documents
Thermo Scientific Chemicals
461760050
5 g
216.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2030-63-9
473.4
2794
C27H22Cl2N4
WDQPAMHFFCXSNU-UHFFFAOYSA-N
CHEBI:3749

Specifications

2030-63-9
97.5% min. (HPLC)
C27H22Cl2N4
WDQPAMHFFCXSNU-UHFFFAOYSA-N
2794
473.4
Clofazimine
219°C
Glass Bottle
Solubility in water: insoluble. Other solubilities: soluble in toluene,slightly soluble in ether,acetone,ethyl acetate
473.4
CHEBI:3749
98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 217-980-2

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only