Learn More
Nickel Electro™ Clifton™ Digital Thermostirrer Water Bath with Clear Plastic Tank and Stainless Steel Shelf
Get maximum observation of samples, water circulation, and temperature stability with the stirred water bath. Designed for immersion of flasks, bottles, racks in a highly stable temperature environment, this Clifton™ Digital Thermostirrer Water Bath with Clear Plastic Tank and Stainless Steel Shelf includes anti-bacterial technology and other unique features as demanded by many clinical and laboratory environments.
Brand:  Nickel Electro™ NE4-8D/CT
 View more versions of this product
Product Code. 12768908
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
These stirred Clifton water baths feature crystal clear tanks and are designed for immersion of flasks, bottles and racks in a stable temperature environment. Durable chemical resistant stainless steel construction with antibacterial paint finish, immersed components are corrosion resistant. Controls are easy to wipe and also have an antibacterial finish. Baths are supplied with a removable stainless steel false base designed for good water circulation and water temperature stability. Features main menu driven data entry, safety cut-out float system for low liquid level protection, factory pre-set two-point calibration and two control options, temperature only or temperature and time.
  • Adaptive PID control allows temperature to be accurately controlled for reproducible conditions
  • Water bath liquid temperature displayed for complete assurance
  • Indication of heating, over temperature alarm, timer and setting set points
  • When a low liquid level is detected a FILL message automatically appears and heating is suspended. Once refilled, heating is restored and temperature control resumes automatically
  • Clear polycarbonate tank, crystal clear, graduated in liters
Specifications
Specifications
Digital Water Bath
CE Approved
Digital LED
NE4-8 D/CT
3 Years
130 x 130 x 150 mm
130 mm
150 mm
130 mm
230 V
50/60 Hz
8 L
5°C to 99°C
Stainless Steel
±0.1°C
265 x 265 x 295 mm
265 mm
295 mm
265 mm
7.5 kg
1250 W
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products