Learn More
Nickel Electro™ Cooling Coil
Nickel Electro™ Cooling Coil immerses into Clifton™ Stirred or Circulating water bath, behind the Stirrer, without affecting bath working area. When connected to a cold water supply it provides counter cooling depending on tap water temperature and flow rate.
Brand:  Nickel Electro™ NE4-CC
Product Code. 12748868
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • This Cooling Coil does not provide forced cooling as provided with DC range of Immersion Dip Coolers.
  • A Lid is recommended in use.
  • Stainless steel
  • For use with cold running water supply.
Specifications
Specifications
Cooling Coil
20 x 10 x 10 cm
20 x 10 x 10 cm