Learn More
Zwiebel™ Plastic Box with Stainless Steel 25kg Weight, F1 with Certificate
Use with weighing instruments of special accuracy class I and high accuracy class II. Zwiebel™ Class F1 Weights are intended for use in the verification or calibration of class F2 weights.
Brand: Zwiebel™ PBI5682N
This item is not returnable.
View return policy
Description
- Class F1 calibration weight
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
Specifications
Box Weight | |
F1 | |
25 kg | |
Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |