missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Plastic Box with Stainless Steel 500g Weight, Class E2

Calibrate weights of lower accuracy classes. Zwiebel™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.

Brand:  Zwiebel™ PBI56813

Product Code. 11820892

  • 397.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Class E2 stainless steel calibration weight
  • In compliance with OIML R111
  • Specifications

    Specifications

    ±0.8 mg
    For use with Weighing Instruments of Special Accuracy Class I
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.