Learn More
Fisherbrand™ Class B Glass Tall Form Measuring Cylinders
Measure and pour liquids with precision with these cylinders. Fisherbrand™ Class B Glass Tall Form Measuring Cylinders are manufactured from calibrated, clear borosilicate glass.
Specifications
Certifications/Compliance | ISO 4788 |
---|---|
Class | B |
Type | Graduated Measuring Cylinder |
Material | Borosilicate Glass |
Autoclavable | Autoclavable |
Description
- Graduated
- Stable hexagonal foot
- Corrosion-free and resistant to thermal shock
Specifications
ISO 4788 | |
Graduated Measuring Cylinder | |
Autoclavable |
B | |
Borosilicate Glass | |
500°C |