Learn More
Honeywell™ Retaining Covers for Prefilter Pads
Retaining Covers for Prefilter Pads are used in conjunction with Honeywell™ cartridges on twin-filter respirators
Brand:  Honeywell™ N750036A
 View more versions of this product
Product Code. 15886635
  • 35.00 € / 1 pair
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Product Type: Retaining Cover
  • Color: White
  • Material: Plastic
  • Requires: Prefilter Pad
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bayonet Gas/Vapor Cartridges on N-Series Respirators
Requires Honeywell™ Prefilter Pads [7506P2 (Particulate, P2)]
Honeywell™ Half-Mask (e.g. N5500™, N7700™) and Full-Face (e.g. N5400™) Air-Purifying Respirators
N750036A
White
Retainer Cover for Prefilter Pads
Spider and Retainer
907.6 g