missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, Powder, Multi-Compendial, J.T. Baker™

Product Code. 15527784
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 77-92-9
Molecular Formula C6H8O7
MDL Number MFCD00011669
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Weight (g/mol) 192.12
PubChem CID 311
ChEBI CHEBI:30769
Formula Weight 192.12
Grade Multi-Compendial
Quantity 2.5 kg
Packaging Poly Bottle
Physical Form Powder
Specific Gravity 1L = 1.67kg
Chemical Name or Material Citric Acid
Show More Show Less
compliance-icons
Product Identifier
  • Citric acid, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Citric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.