missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, Honeywell Fluka™
BIOPHARMA

Tested according to European Pharmacopoeia

160.11 € - 713.01 €

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.138
MDL Number MFCD00149972
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
PubChem CID 22230
ChEBI CHEBI:31404
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15602920
BIOPHARMA
View Documents Promotion Details
Honeywell-Fluka
27491-1KG
1 kg
160.11 €
1kg
Please to purchase this item. Need a web account? Register with us today!
15612920
View Documents Promotion Details
Honeywell-Fluka
27491-25KG
25 kg
713.01 €
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

5949-29-1
210.138
YASYEJJMZJALEJ-UHFFFAOYSA-N
22230
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
C6H10O8
MFCD00149972
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
CHEBI:31404
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O

Specifications

5949-29-1
HOC(COOH)(CH2COOH)2 · H2O
NONH for all modes of transport
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
YASYEJJMZJALEJ-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
22230
210.14g/mol
Citric Acid Monohydrate, Tested according to Ph.Eur.
C6H10O8
MFCD00149972
4018641
Soluble in ethyl acetate (5.28g/100g), Amyl Acetate(5.98g/100g), diethyl ether(2.17g/100g), methanol(197g/100g), chloroform(0.007g/100 g)
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
210.138
CHEBI:31404
173°C (343.4°F)
Videos
Safety and Handling

Safety and Handling

P305 + P351 + P338

H319

missing translation for 'einecsNumber' : 201-069-1

missing translation for 'rtecsNumber' : GE7810000

Product Identifier
  • Citric acid monohydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Citric acid monohydrate 5

SDS
Documents

Documents

Product Certifications