missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid Monohydrate, Extra Pure, SLR, Granular, Fisher Chemical™
21.90 € - 110.00 €
Chemical Identifiers
CAS | 5949-29-1 |
---|---|
Molecular Formula | C6H10O8 |
Molecular Weight (g/mol) | 210.14 |
MDL Number | 149972 |
InChI Key | YASYEJJMZJALEJ-UHFFFAOYSA-N |
IUPAC Name | 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate |
SMILES | O.OC(=O)CC(O)(CC(O)=O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10233470
|
Fisher Chemical
C/6160/53 |
500 g | Plastic powder jar |
21.90 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10745901
|
Fisher Chemical
C/6160/60 |
1 kg | Plastic powder jar |
47.40 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10161850
|
Fisher Chemical
C/6160/62 |
2.5 kg | Plastic powder jar |
68.40 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10429170
|
Fisher Chemical
C/6160/65 |
5 kg | Plastic keg |
110.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
5949-29-1 | |
210.14 | |
YASYEJJMZJALEJ-UHFFFAOYSA-N | |
O.OC(=O)CC(O)(CC(O)=O)C(O)=O |
C6H10O8 | |
149972 | |
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate |
Specifications
1.8 | |
135°C | |
C6H10O8 | |
YASYEJJMZJALEJ-UHFFFAOYSA-N | |
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate | |
210.15g/mol |
5949-29-1 | |
White | |
149972 | |
O.OC(=O)CC(O)(CC(O)=O)C(O)=O | |
210.14 | |
Solid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Citric acid monohydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Citric acid monohydrate, Citric acid