Learn More
Citric acid monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
27.03 € - 120.60 €
Chemical Identifiers
CAS | 5949-29-1 |
---|---|
Molecular Formula | C6H8O7·H2O |
Molecular Weight (g/mol) | 210.15 |
InChI Key | YASYEJJMZJALEJ-UHFFFAOYSA-N |
Synonym | citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp |
PubChem CID | 22230 |
ChEBI | CHEBI:31404 |
IUPAC Name | 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate |
SMILES | C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12095350
|
Thermo Scientific Acros
385852500 |
250 g | Plastic bottle |
27.03 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12075350
|
Thermo Scientific Acros
385850010 |
1 kg | Plastic bottle |
66.53 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12085350
|
Thermo Scientific Acros
385850025 |
2.5 kg | Plastic bottle |
120.60 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5949-29-1 | |
210.15 | |
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp | |
CHEBI:31404 | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O |
C6H8O7·H2O | |
YASYEJJMZJALEJ-UHFFFAOYSA-N | |
22230 | |
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate |
Specifications
5949-29-1 | |
174°C | |
99+% | |
HO2CCH2C(OH)(CO2H)CH2CO2H·H2O | |
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp | |
YASYEJJMZJALEJ-UHFFFAOYSA-N | |
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate | |
22230 | |
210.15 | |
ACS Reagent | |
Citric acid monohydrate |
135.0°C to 152.0°C | |
0.02% max | |
C6H8O7·H2O | |
15,2325 | |
Solubility in water: 676g/L (25°C). Other solubilities: 2.17g/100g ether - 0.007g/100g chloroform, 15.43g/100g amyl alcohol - 5.98g/100g amyl acetate, 5.28g/100g ethyl acetate - 197g/100g methanol, 62.8g/100g propanol | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O | |
210.15 | |
CHEBI:31404 | |
99+% | |
0.005% max |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact len
GHS Signal Word: Warning
- Citric acid monohydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.