missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Citalopram hydrobromide
51.76 € - 161.81 €
Chemical Identifiers
CAS | 59729-32-7 |
---|---|
Molecular Formula | C20H21FN2O·HBr |
Molecular Weight (g/mol) | 405.3 |
InChI Key | WIHMBLDNRMIGDW-UHFFFAOYSA-N |
Synonym | citalopram hydrobromide, citalopram hbr, cipramil, celexa, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide, elopram, seropram, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide, citalopram hydrobromide usan |
PubChem CID | 77995 |
IUPAC Name | 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrobromide |
SMILES | CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15722987
|
Thermo Scientific Chemicals
462310010 |
1 g |
51.76 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15732987
|
Thermo Scientific Chemicals
462310050 |
5 g |
161.81 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
59729-32-7 | |
405.3 | |
citalopram hydrobromide, citalopram hbr, cipramil, celexa, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide, elopram, seropram, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide, citalopram hydrobromide usan | |
1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrobromide |
C20H21FN2O·HBr | |
WIHMBLDNRMIGDW-UHFFFAOYSA-N | |
77995 | |
CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.Br |
Specifications
59729-32-7 | |
C20H21FN2O·HBr | |
(1% in ethanol) Clear colorless | |
CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.Br | |
405.3 | |
405.3 |
185.0°C to 188.0°C | |
citalopram hydrobromide, citalopram hbr, cipramil, celexa, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide, elopram, seropram, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide, citalopram hydrobromide usan | |
WIHMBLDNRMIGDW-UHFFFAOYSA-N | |
1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrobromide | |
77995 | |
Citalopram hydrobromide |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 261-890-6
RUO – Research Use Only