Learn More
cis-4-Hexen-1-ol, 97%, Thermo Scientific Chemicals
39.00 € - 141.00 €
Chemical Identifiers
CAS | 928-91-6 |
---|---|
Molecular Formula | C6H12O |
Molecular Weight (g/mol) | 100.16 |
MDL Number | MFCD00148974,MFCD00009713 |
InChI Key | VTIODUHBZHNXFP-IHWYPQMZSA-N |
Synonym | cis-4-hexen-1-ol, z-hex-4-en-1-ol, z-4-hexen-1-ol, 4z-hex-4-en-1-ol, 4-hexen-1-ol, z, unii-780wma6j84, e-4-hexenol, cis-1-hydroxy-4-hexene, cis-4-hexene-1-ol, 4z-4-hexen-1-ol # |
PubChem CID | 5365589 |
IUPAC Name | (Z)-hex-4-en-1-ol |
SMILES | C\C=C/CCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11419536
|
Thermo Scientific Alfa Aesar
A19266.06 |
5 g |
39.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429536
|
Thermo Scientific Alfa Aesar
A19266.14 |
25 g |
141.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
928-91-6 | |
100.16 | |
VTIODUHBZHNXFP-IHWYPQMZSA-N | |
5365589 | |
C\C=C/CCCO |
C6H12O | |
MFCD00148974,MFCD00009713 | |
cis-4-hexen-1-ol, z-hex-4-en-1-ol, z-4-hexen-1-ol, 4z-hex-4-en-1-ol, 4-hexen-1-ol, z, unii-780wma6j84, e-4-hexenol, cis-1-hydroxy-4-hexene, cis-4-hexene-1-ol, 4z-4-hexen-1-ol # | |
(Z)-hex-4-en-1-ol |
Specifications
928-91-6 | |
65°C (149°F) | |
C6H12O | |
UN1987 | |
VTIODUHBZHNXFP-IHWYPQMZSA-N | |
(Z)-hex-4-en-1-ol | |
5365589 | |
97% |
158°C | |
Pleasant | |
MFCD00148974,MFCD00009713 | |
cis-4-hexen-1-ol, z-hex-4-en-1-ol, z-4-hexen-1-ol, 4z-hex-4-en-1-ol, 4-hexen-1-ol, z, unii-780wma6j84, e-4-hexenol, cis-1-hydroxy-4-hexene, cis-4-hexene-1-ol, 4z-4-hexen-1-ol # | |
C\C=C/CCCO | |
100.16 | |
100.16 | |
cis-4-Hexen-1-ol |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only