Learn More
Cinnamyl alcohol, 98% trans, Thermo Scientific Chemicals
23.00 € - 45.70 €
Chemical Identifiers
CAS | 104-54-1 |
---|---|
Molecular Formula | C9H10O |
Molecular Weight (g/mol) | 134.18 |
MDL Number | MFCD00002921 |
InChI Key | OOCCDEMITAIZTP-QPJJXVBHSA-N |
Synonym | cinnamyl alcohol, cinnamic alcohol, 3-phenyl-2-propen-1-ol, 3-phenylprop-2-en-1-ol, zimtalcohol, styryl carbinol, e-3-phenylprop-2-en-1-ol, 3-phenylallyl alcohol, trans-cinnamyl alcohol, styryl alcohol |
PubChem CID | 5315892 |
ChEBI | CHEBI:33227 |
IUPAC Name | (E)-3-phenylprop-2-en-1-ol |
SMILES | C1=CC=C(C=C1)C=CCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10746361
|
Thermo Scientific Acros
148520050 |
5 g | Glass bottle |
23.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10755041
|
Thermo Scientific Acros
148521000 |
100 g | Glass bottle |
27.60 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10645772
|
Thermo Scientific Acros
148522500 |
250 g | Glass bottle |
27.30 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10316890
|
Thermo Scientific Acros
148525000 |
500 g | Glass bottle |
45.70 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
104-54-1 | |
134.18 | |
OOCCDEMITAIZTP-QPJJXVBHSA-N | |
5315892 | |
(E)-3-phenylprop-2-en-1-ol |
C9H10O | |
MFCD00002921 | |
cinnamyl alcohol, cinnamic alcohol, 3-phenyl-2-propen-1-ol, 3-phenylprop-2-en-1-ol, zimtalcohol, styryl carbinol, e-3-phenylprop-2-en-1-ol, 3-phenylallyl alcohol, trans-cinnamyl alcohol, styryl alcohol | |
CHEBI:33227 | |
C1=CC=C(C=C1)C=CCO |
Specifications
104-54-1 | |
White | |
258.0°C | |
Authentic | |
C6H5CH=CHCH2OH | |
06, 570 | |
1.04 | |
Solubility in water: 1.8g/L (20°C). Other solubilities: soluble in glycerol, methanol, clearly soluble in 3 volumes 50% alcohol, freely soluble in alcohol, ether, and other, common organic solvents | |
C1=CC=C(C=C1)C=CCO | |
134.18 | |
CHEBI:33227 | |
98% trans | |
Cinnamyl alcohol, 98% |
31.0°C to 35.0°C | |
1.0400g/mL | |
126°C | |
C9H10O | |
MFCD00002921 | |
15, 2303 | |
cinnamyl alcohol, cinnamic alcohol, 3-phenyl-2-propen-1-ol, 3-phenylprop-2-en-1-ol, zimtalcohol, styryl carbinol, e-3-phenylprop-2-en-1-ol, 3-phenylallyl alcohol, trans-cinnamyl alcohol, styryl alcohol | |
OOCCDEMITAIZTP-QPJJXVBHSA-N | |
(E)-3-phenylprop-2-en-1-ol | |
5315892 | |
134.18 | |
Fused Low Melting Crystalline Solid |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Contaminated work clothing should not be allowed out of the workplace.
Avoid breathing dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-212-3
missing translation for 'rtecsNumber' : GE2200000
missing translation for 'tsca' : TSCA
- Cinnamyl alcohol
Signal Word
- Warning
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only